2'-Chloro-6'-methyl-2,2,2-trifluoroacetophenone ≥97%

Supplier: Matrix Scientific

086935-1G 086935-2G 086935-5G
102851-512EA 552.31 USD
102851-512 102851-514 102851-516
2'-Chloro-6'-methyl-2,2,2-trifluoroacetophenone ≥97%
2'-Chloro-6'-methyl-2,2,2-trifluoroacetophenone

Hazard: Irritant

Formula: C₉H₆ClF₃O
MDL Number: MFCD02260846
CAS Number: 886371-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR