2',3'-Dichloro-2,2,2-trifluoroacetophenone ≥97%

Supplier: Matrix Scientific

086933-1G 086933-2G 086933-5G
102851-500EA 347.69 USD
102851-500 102851-502 102851-504
2',3'-Dichloro-2,2,2-trifluoroacetophenone ≥97%
2',3'-Dichloro-2,2,2-trifluoroacetophenone

Hazard: Irritant

Formula: C₈H₃Cl₂F₃O
MDL Number: MFCD02260843
CAS Number: 886371-11-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR