2,2,2,2',4'-Pentafluoroacetophenone ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(2,4-Difluorophenyl)-2,2,2-trifluoroethanone

086932-1G 086932-2G 086932-5G
102851-494EA 347.69 USD
102851-494 102851-496 102851-498
2,2,2,2',4'-Pentafluoroacetophenone ≥97%
2,2,2,2',4'-Pentafluoroacetophenone

Hazard: Irritant

Formula: C₈H₃F₅O
MW: 210.10 g/mol
MDL Number: MFCD02260842
CAS Number: 886371-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR