4'-Chloro-2'-fluoro-2,2,2-trifluoroacetophenone ≥97%
Supplier: Matrix Scientific
Synonyms:
1-(4-Chloro-2-fluoro-phenyl)-2,2,2-trifluoro-ethanone
086931-1G
086931-2G
086931-5G
102851-488EA
872.31
USD
102851-488
102851-490
102851-492
4'-Chloro-2'-fluoro-2,2,2-trifluoroacetophenone ≥97%
4'-Chloro-2'-fluoro-2,2,2-trifluoroacetophenone
Hazard: Irritant
Formula:
C₈H₃ClF₄O MW: 226.56 g/mol |
MDL Number:
MFCD02260841 CAS Number: 886370-99-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...