4'-Methoxy-3'-methyl-2,2,2-trifluoroacetophenone ≥97%

Supplier: Matrix Scientific

086929-1G 086929-2G 086929-5G
102851-476EA 347.69 USD
102851-476 102851-478 102851-480
4'-Methoxy-3'-methyl-2,2,2-trifluoroacetophenone ≥97%
4'-Methoxy-3'-methyl-2,2,2-trifluoroacetophenone

Hazard: Irritant

Formula: C₁₀H₉F₃O₂
MDL Number: MFCD02260834
CAS Number: 75822-11-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR