3'-Chloro-2,2,2,5'-tetrafluoroacetophenone ≥97%

Supplier: Matrix Scientific

086928-1G 086928-2G 086928-5G
102851-470EA 552.31 USD
102851-470 102851-472 102851-474
3'-Chloro-2,2,2,5'-tetrafluoroacetophenone ≥97%
3'-Chloro-2,2,2,5'-tetrafluoroacetophenone

Hazard: Irritant

Formula: C₈H₃ClF₄O
MDL Number: MFCD02260831
CAS Number: 845823-07-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR