4'-Chloro-3'-methyl-2,2,2-trifluoroacetophenone ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(4-Chloro-3-methylphenyl)-2,2,2-trifluoroethanone

086926-1G 086926-2G 086926-5G
102851-458EA 552.31 USD
102851-458 102851-460 102851-462
4'-Chloro-3'-methyl-2,2,2-trifluoroacetophenone ≥97%
4'-Chloro-3'-methyl-2,2,2-trifluoroacetophenone

Hazard: Irritant

Formula: C₉H₆ClF₃O
MDL Number: MFCD02260828
CAS Number: 286017-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR