3'-Bromo-2,2,2-trifluoroacetophenone ≥97%

Supplier: Matrix Scientific

086925-1G 086925-2G 086925-5G
102851-452EA 552.31 USD
102851-452 102851-454 102851-456
3'-Bromo-2,2,2-trifluoroacetophenone ≥97%
3'-Bromo-2,2,2-trifluoroacetophenone

Hazard: Irritant/Refrigerate

Formula: C₈H₄BrF₃O
MDL Number: MFCD00511232
CAS Number: 655-26-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR