4'-Phenoxy-2,2,2-trifluoroacetophenone ≥97%

Supplier: Matrix Scientific

086924-1G 086924-2G 086924-5G
102851-446EA 552.31 USD
102851-446 102851-448 102851-450
4'-Phenoxy-2,2,2-trifluoroacetophenone ≥97%
4'-Phenoxy-2,2,2-trifluoroacetophenone

Hazard: Irritant

Formula: C₁₄H₉F₃O₂
MDL Number: MFCD00042392
CAS Number: 70783-32-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR