4'-Phenyl-2,2,2-trifluoroacetophenone ≥97%

Supplier: Matrix Scientific

086923-1G 086923-2G 086923-5G
102851-440EA 552.31 USD
102851-440 102851-442 102851-444
4'-Phenyl-2,2,2-trifluoroacetophenone ≥97%
4'-Phenyl-2,2,2-trifluoroacetophenone

Hazard: Irritant

Formula: C₁₄H₉F₃O
MDL Number: MFCD01320014
CAS Number: 2369-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR