2'-Phenyl-2,2,2-trifluoroacetophenone ≥97%

Supplier: Matrix Scientific

Synonyms: 1-([1,1'-Biphenyl]-2-yl)-2,2,2-trifluoroethanone

086922-1G 086922-2G 086922-5G
102851-434EA 552.31 USD
102851-434 102851-436 102851-438
2'-Phenyl-2,2,2-trifluoroacetophenone ≥97%
2'-Phenyl-2,2,2-trifluoroacetophenone

Hazard: Irritant

Formula: C₁₄H₉F₃O
MDL Number: MFCD01320013
CAS Number: 302912-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR