1-(4-Methylnaphthyl) trifluoromethyl ketone ≥97%
Supplier: Matrix Scientific
086920-1G
086920-2G
086920-5G
102851-422EA
872.31
USD
102851-422
102851-424
102851-426
1-(4-Methylnaphthyl) trifluoromethyl ketone ≥97%
1-(4-Methylnaphthyl) trifluoromethyl ketone
Hazard: Irritant
Formula:
C₁₃H₉F₃O |
MDL Number:
MFCD01320010 CAS Number: 886370-33-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...