4'-(Dimethylamino)-2,2,2-trifluoroacetophenone ≥97%

Supplier: Matrix Scientific

086917-1G 086917-2G 086917-5G
102851-404EA 347.69 USD
102851-404 102851-406 102851-408
4'-(Dimethylamino)-2,2,2-trifluoroacetophenone ≥97%
4'-(Dimethylamino)-2,2,2-trifluoroacetophenone

Hazard: Irritant

Formula: C₁₀H₁₀F₃NO
MW: 217.19 g/mol
Melting Pt: 74…75 °C
MDL Number: MFCD00099468
CAS Number: 2396-05-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR