2'-Methyl-2,2,2,3'-tetrafluoroacetophenone ≥97%

Supplier: Matrix Scientific

086916-1G 086916-2G 086916-5G
102851-398EA 347.69 USD
102851-398 102851-400 102851-402
2'-Methyl-2,2,2,3'-tetrafluoroacetophenone ≥97%
2'-Methyl-2,2,2,3'-tetrafluoroacetophenone

Hazard: Irritant

Formula: C₉H₆F₄O
MDL Number: MFCD01320002
CAS Number: 886370-05-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR