2'-Methyl-2,2,2,4'-tetrafluoroacetophenone ≥97%

Supplier: Matrix Scientific

Synonyms: 2,2,2-Trifluoro-1-(4-fluoro-2-methylphenyl)ethanone

086915-1G 086915-2G 086915-5G
102851-392EA 552.31 USD
102851-392 102851-394 102851-396
2'-Methyl-2,2,2,4'-tetrafluoroacetophenone ≥97%
2'-Methyl-2,2,2,4'-tetrafluoroacetophenone

Hazard: Irritant

Formula: C₉H₆F₄O
MW: 206.14 g/mol
MDL Number: MFCD01320001
CAS Number: 886370-02-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR