2'-Methyl-2,2,2,5'-tetrafluoroacetophenone ≥97%
Supplier: Matrix Scientific
086914-1G
086914-2G
086914-5G
102851-386EA
552.31
USD
102851-386
102851-388
102851-390
2'-Methyl-2,2,2,5'-tetrafluoroacetophenone ≥97%
2'-Methyl-2,2,2,5'-tetrafluoroacetophenone
Hazard: Irritant
Formula:
C₉H₆F₄O MW: 206.14 g/mol |
MDL Number:
MFCD01320000 CAS Number: 886369-93-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...