3',4'-Dimethoxy-2,2,2-trifluoroacetophenone ≥97%

Supplier: Matrix Scientific

086909-1G 086909-2G 086909-5G
102851-356EA 552.31 USD
102851-356 102851-358 102851-360
3',4'-Dimethoxy-2,2,2-trifluoroacetophenone ≥97%
3',4'-Dimethoxy-2,2,2-trifluoroacetophenone

Hazard: Irritant

Formula: C₁₀H₉F₃O₃
MDL Number: MFCD01319991
CAS Number: 300374-83-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR