3',4'-Dimethyl-2,2,2-trifluoroacetophenone ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(3,4-Dimethylphenyl)-2,2,2-trifluoroethanone

086907-1G 086907-2G 086907-5G
102851-344EA 520 USD
102851-344 102851-346 102851-348
3',4'-Dimethyl-2,2,2-trifluoroacetophenone ≥97%
3',4'-Dimethyl-2,2,2-trifluoroacetophenone

Hazard: Refrigerate

Formula: C₁₀H₉F₃O
MDL Number: MFCD01319987
CAS Number: 75833-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR