2',6'-Dimethyl-2,2,2-trifluoroacetophenone ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(2,6-Dimethylphenyl)-2,2,2-trifluoroethanone

086906-1G 086906-2G 086906-5G
102851-338EA 347.69 USD
102851-338 102851-340 102851-342
2',6'-Dimethyl-2,2,2-trifluoroacetophenone ≥97%
2',6'-Dimethyl-2,2,2-trifluoroacetophenone

Hazard: Refrigerate

Formula: C₁₀H₉F₃O
MDL Number: MFCD01319986
CAS Number: 121456-63-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR