4'-tert-Butyl-2,2,2-trifluoroacetophenone ≥97%
Supplier: Matrix Scientific
086903-1G
086903-2G
086903-5G
102851-320EA
347.69
USD
102851-320
102851-322
102851-324
4'-tert-Butyl-2,2,2-trifluoroacetophenone ≥97%
4'-tert-Butyl-2,2,2-trifluoroacetophenone
Hazard: Irritant
Formula:
C₁₂H₁₃F₃O |
MDL Number:
MFCD01319979 CAS Number: 73471-97-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...