2'-(Methylthio)-2,2,2-trifluoroacetophenone ≥97%

Supplier: Matrix Scientific

086901-1G 086901-2G 086901-5G
102851-308EA 347.69 USD
102851-308 102851-310 102851-312
2'-(Methylthio)-2,2,2-trifluoroacetophenone ≥97%
2'-(Methylthio)-2,2,2-trifluoroacetophenone

Hazard: Irritant

Formula: C₉H₇F₃OS
MDL Number: MFCD01319975
CAS Number: 166831-65-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR