2,2,2,2'-Tetrafluoroacetophenone ≥97%

Supplier: Matrix Scientific

Synonyms: 2,2,2-Trifluoro-1-(2-fluorophenyl)ethanone

086900-1G 086900-2G 086900-5G
102851-302EA 2043.08 USD
102851-302 102851-304 102851-306
2,2,2,2'-Tetrafluoroacetophenone ≥97%
2,2,2,2'-Tetrafluoroacetophenone

Hazard: Refrigerate

Formula: C₈H₄F₄O
MW: 192.1104 g/mol
MDL Number: MFCD00040099
CAS Number: 124004-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR