2'-Chloro-2,2,2-trifluoroacetophenone ≥97%

Supplier: Matrix Scientific

086898-1G 086898-2G 086898-5G
102851-290EA 520 USD
102851-290 102851-292 102851-294
2'-Chloro-2,2,2-trifluoroacetophenone ≥97%
2'-Chloro-2,2,2-trifluoroacetophenone

Hazard: Refrigerate

Formula: C₈H₄ClF₃O
MDL Number: MFCD02260827
CAS Number: 5860-95-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR