4'-Carboethoxy-2,2,2-trifluoroacetophenone ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 4-(2,2,2-Trifluoroacetyl)benzoate

086897-1G 086897-2G 086897-5G
102851-284EA 775.38 USD
102851-284 102851-286 102851-288
4'-Carboethoxy-2,2,2-trifluoroacetophenone ≥97%
4'-Carboethoxy-2,2,2-trifluoroacetophenone

Hazard: Irritant

Formula: C₁₁H₉F₃O₃
MW: 246.19 g/mol
MDL Number: MFCD01319974
CAS Number: 898787-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR