3'-Carboethoxy-2,2,2-trifluoroacetophenone ≥97%

Supplier: Matrix Scientific

086896-1G 086896-2G 086896-5G
102851-278EA 872.31 USD
102851-278 102851-280 102851-282
3'-Carboethoxy-2,2,2-trifluoroacetophenone ≥97%
3'-Carboethoxy-2,2,2-trifluoroacetophenone

Hazard: Irritant/Refrigerate

Formula: C₁₁H₉F₃O₃
MDL Number: MFCD01319973
CAS Number: 898787-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR