3'-Cyano-2,2,2-trifluoroacetophenone ≥97%

Supplier: Matrix Scientific

086893-1G 086893-2G 086893-5G
102851-260EA 920 USD
102851-260 102851-262 102851-264
3'-Cyano-2,2,2-trifluoroacetophenone ≥97%
3'-Cyano-2,2,2-trifluoroacetophenone

Hazard: Irritant

Formula: C₉H₄F₃NO
Melting Pt: 72…75 °C
MDL Number: MFCD01319970
CAS Number: 23568-85-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR