3'-Methoxy-2,2,2-trifluoroacetophenone ≥97%
Supplier: Matrix Scientific
086891-1G
086891-2G
086891-5G
102851-248EA
246.15
USD
102851-248
102851-250
102851-252
3'-Methoxy-2,2,2-trifluoroacetophenone ≥97%
3'-Methoxy-2,2,2-trifluoroacetophenone
Hazard: Refrigerate
Formula:
C₉H₇F₃O₂ |
MDL Number:
MFCD00461895 CAS Number: 30724-22-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...