3'-Methyl-2,2,2-trifluoroacetophenone ≥97%

Supplier: Matrix Scientific

Synonyms: 2,2,2-Trifluoro-1-(m-tolyl)ethanone

086890-1G 086890-2G 086890-5G
102851-242EA 347.69 USD
102851-242 102851-244 102851-246
3'-Methyl-2,2,2-trifluoroacetophenone ≥97%
3'-Methyl-2,2,2-trifluoroacetophenone

Hazard: Irritant

Formula: C₉H₇F₃O
MW: 188.1465 g/mol
MDL Number: MFCD00461893
CAS Number: 1736-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR