2'-Methyl-2,2,2-trifluoroacetophenone ≥97%
Supplier: Matrix Scientific
086889-1G
086889-2G
086889-5G
102851-236EA
453.85
USD
102851-236
102851-238
102851-240
2'-Methyl-2,2,2-trifluoroacetophenone ≥97%
2'-Methyl-2,2,2-trifluoroacetophenone
Hazard: Refrigerate
Formula:
C₉H₇F₃O |
CAS Number:
341-39-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...