3,5-Dimethylbenzoylacetonitrile ≥97%

Supplier: Matrix Scientific

086887-1G 086887-2G 086887-5G
102851-224EA 387.69 USD
102851-224 102851-226 102851-228
3,5-Dimethylbenzoylacetonitrile ≥97%
3,5-Dimethylbenzoylacetonitrile

Hazard: Irritant

Formula: C₁₁H₁₁NO
MDL Number: MFCD00158942
CAS Number: 85692-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR