2,4,5-Trifluorobenzoylacetonitrile ≥97%

Supplier: Matrix Scientific

086886-1G 086886-2G 086886-5G
102851-218EA 592.31 USD
102851-218 102851-220 102851-222
2,4,5-Trifluorobenzoylacetonitrile ≥97%
2,4,5-Trifluorobenzoylacetonitrile

Hazard: Irritant

Formula: C₉H₄F₃NO
MDL Number: MFCD02260821
CAS Number: 142501-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR