2-Naphthoylacetonitrile ≥97%

Supplier: Matrix Scientific

086884-1G 086884-2G 086884-5G
102851-206EA 592.31 USD
102851-206 102851-208 102851-210
2-Naphthoylacetonitrile ≥97%
2-Naphthoylacetonitrile

Hazard: Irritant

Formula: C₁₃H₉NO
MDL Number: MFCD02260808
CAS Number: 613-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR