2,4-Difluorobenzoylacetonitrile ≥97%

Supplier: Matrix Scientific

086882-1G 086882-2G 086882-5G
102851-194EA 592.31 USD
102851-194 102851-196 102851-198
2,4-Difluorobenzoylacetonitrile ≥97%
2,4-Difluorobenzoylacetonitrile

Hazard: Irritant

Formula: C₉H₅F₂NO
MDL Number: MFCD02260804
CAS Number: 71682-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR