2-Methylbenzoylacetonitrile ≥97%
Supplier: Matrix Scientific
Synonyms:
3-(2-Methylphenyl)-3-oxopropanenitrile, 3-Oxo-3-(o-tolyl)propanenitrile
086881-1G
086881-2G
086881-5G
102851-188EA
592.31
USD
102851-188
102851-190
102851-192
2-Methylbenzoylacetonitrile ≥97%
2-Methylbenzoylacetonitrile
Hazard: Irritant
Formula:
C₁₀H₉NO MW: 159.19 g/mol |
MDL Number:
MFCD02260802 CAS Number: 35276-81-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...