3,5-Dimethyl-4-methoxybenzoylacetonitrile ≥97%

Supplier: Matrix Scientific

086879-1G 086879-5G
102851-176EA 387.69 USD
102851-176 102851-180
3,5-Dimethyl-4-methoxybenzoylacetonitrile ≥97%
3,5-Dimethyl-4-methoxybenzoylacetonitrile

Hazard: Irritant

Formula: C₁₂H₁₃NO₂
MDL Number: MFCD02260791
CAS Number: 884504-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR