3,4-Dimethylbenzoylacetonitrile ≥97%

Supplier: Matrix Scientific

Synonyms: 3-(3,4-Dimethylphenyl)-3-oxopropanenitrile

086878-1G 086878-2G 086878-5G
102851-170EA 387.69 USD
102851-170 102851-172 102851-174
3,4-Dimethylbenzoylacetonitrile ≥97%
3,4-Dimethylbenzoylacetonitrile

Hazard: Irritant

Formula: C₁₁H₁₁NO
MW: 173.22 g/mol
MDL Number: MFCD02260786
CAS Number: 884504-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR