3-Fluorobenzoylacetonitrile ≥97%

Supplier: Matrix Scientific

Synonyms: 3-(3-Fluorophenyl)-3-oxopropanenitrile

086877-1G 086877-2G 086877-5G
102851-164EA 387.69 USD
102851-164 102851-166 102851-168
3-Fluorobenzoylacetonitrile ≥97%
3-Fluorobenzoylacetonitrile

Hazard: Irritant

Formula: C₉H₆FNO
MW: 163.15 g/mol
MDL Number: MFCD02260777
CAS Number: 21667-61-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR