1-Chloro-3-(3,4-dimethoxyphenyl)-3-oxopropane ≥95%

Supplier: Matrix Scientific

086875-1G 086875-2G 086875-5G
102851-152EA 340 USD
102851-152 102851-154 102851-156
1-Chloro-3-(3,4-dimethoxyphenyl)-3-oxopropane ≥95%
1-Chloro-3-(3,4-dimethoxyphenyl)-3-oxopropane

Hazard: Irritant

Formula: C₁₁H₁₃ClO₃
MDL Number: MFCD00510412
CAS Number: 4693-38-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR