(3-Bromophenyl)acetone ≥97%

Supplier: Matrix Scientific

086874-1G 086874-2G 086874-5G
102851-146EA 592.31 USD
102851-146 102851-148 102851-150
(3-Bromophenyl)acetone ≥97%
(3-Bromophenyl)acetone

Hazard: Irritant

Formula: C₉H₉BrO
MDL Number: MFCD00210400
CAS Number: 21906-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR