5-(4-Methoxyphenyl)-5-oxovaleronitrile ≥97%

Supplier: Matrix Scientific

086871-1G 086871-2G 086871-5G
102851-128EA 347.69 USD
102851-128 102851-130 102851-132
5-(4-Methoxyphenyl)-5-oxovaleronitrile ≥97%
5-(4-Methoxyphenyl)-5-oxovaleronitrile

Hazard: Irritant

Formula: C₁₂H₁₃NO₂
MDL Number: MFCD00448777
CAS Number: 26823-02-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR