6-Chloro-1-(4-methoxyphenyl)-1-oxohexane ≥97%
Supplier: Matrix Scientific
086868-1G
086868-2G
086868-5G
102851-110EA
347.69
USD
102851-110
102851-112
102851-114
6-Chloro-1-(4-methoxyphenyl)-1-oxohexane ≥97%
6-Chloro-1-(4-methoxyphenyl)-1-oxohexane
Hazard: Irritant
Formula:
C₁₃H₁₇ClO₂ MW: 240.73 g/mol |
MDL Number:
MFCD07700094 CAS Number: 278619-91-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...