5-Chloro-1-(4-methoxyphenyl)-1-oxopentane ≥97%

Supplier: Matrix Scientific

086867-1G 086867-2G 086867-5G
102851-104EA 347.69 USD
102851-104 102851-106 102851-108
5-Chloro-1-(4-methoxyphenyl)-1-oxopentane ≥97%
5-Chloro-1-(4-methoxyphenyl)-1-oxopentane

Hazard: Irritant

Formula: C₁₂H₁₅ClO₂
MDL Number: MFCD00517502
CAS Number: 949-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR