4-Chloro-1-(4-cyanophenyl)-1-oxobutane ≥95%
Supplier: Matrix Scientific
086866-1G
086866-2G
086866-5G
102851-098EA
1191.4
USD
102851-098
102851-100
102851-102
4-Chloro-1-(4-cyanophenyl)-1-oxobutane ≥95%
4-Chloro-1-(4-cyanophenyl)-1-oxobutane
Hazard: Irritant
Formula:
C₁₁H₁₀ClNO |
MDL Number:
MFCD07700076 CAS Number: 188851-50-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...