4-Chloro-1-oxo-1-(4-trifluoromethoxyphenyl)butane ≥95%

Supplier: Matrix Scientific

086865-1G 086865-2G 086865-5G
102851-092EA 347.69 USD
102851-092 102851-094 102851-096
4-Chloro-1-oxo-1-(4-trifluoromethoxyphenyl)butane ≥95%
4-Chloro-1-oxo-1-(4-trifluoromethoxyphenyl)butane

Hazard: Irritant

Formula: C₁₁H₁₀ClF₃O₂
MDL Number: MFCD07700070
CAS Number: 898761-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR