Cyclopropyl-4-(1,3-dioxolan-2-yl)phenyl ketone ≥97%

Supplier: Matrix Scientific

086862-1G 086862-2G 086862-5G
102851-074EA 672.31 USD
102851-074 102851-076 102851-078
Cyclopropyl-4-(1,3-dioxolan-2-yl)phenyl ketone ≥97%
Cyclopropyl-4-(1,3-dioxolan-2-yl)phenyl ketone

Hazard: Irritant

Formula: C₁₃H₁₄O₃
MDL Number: MFCD07700044
CAS Number: 898760-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR