4'-(1,3-Dioxolan-2-yl)-2-fluorobenzophenone ≥97%

Supplier: Matrix Scientific

086861-1G 086861-2G 086861-5G
102851-068EA 672.31 USD
102851-068 102851-070 102851-072
4'-(1,3-Dioxolan-2-yl)-2-fluorobenzophenone ≥97%

Hazard: Irritant

Formula: C₁₆H₁₃FO₃
MDL Number: MFCD07700027
CAS Number: 898760-46-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR