3-(1,3-Dioxolan-2-yl)-4'-methoxybenzophenone ≥97%

Supplier: Matrix Scientific

086860-1G 086860-2G 086860-5G
102851-062EA 672.31 USD
102851-062 102851-064 102851-066
3-(1,3-Dioxolan-2-yl)-4'-methoxybenzophenone ≥97%
3-(1,3-Dioxolan-2-yl)-4'-methoxybenzophenone

Hazard: Irritant

Formula: C₁₇H₁₆O₄
MDL Number: MFCD07699951
CAS Number: 898778-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR