2'-Methyl-3-(3,4,5-trifluorophenyl)propiophenone ≥97%

Supplier: Matrix Scientific

086859-1G 086859-2G 086859-5G
102851-056EA 672.31 USD
102851-056 102851-058 102851-060
2'-Methyl-3-(3,4,5-trifluorophenyl)propiophenone ≥97%
2'-Methyl-3-(3,4,5-trifluorophenyl)propiophenone

Hazard: Irritant

Formula: C₁₆H₁₃F₃O
MDL Number: MFCD03844024
CAS Number: 898777-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR