3-(4-Chlorophenyl)-4'-fluoropropiophenone ≥97%

Supplier: Matrix Scientific

086858-1G 086858-2G 086858-5G
102851-050EA 672.31 USD
102851-050 102851-052 102851-054
3-(4-Chlorophenyl)-4'-fluoropropiophenone ≥97%
3-(4-Chlorophenyl)-4'-fluoropropiophenone

Hazard: Irritant

Formula: C₁₅H₁₂ClFO
MDL Number: MFCD03843471
CAS Number: 56201-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR