4'-Bromo-3-(4-chlorophenyl)propiophenone ≥97%

Supplier: Matrix Scientific

086857-1G 086857-2G 086857-5G
102851-044EA 672.31 USD
102851-044 102851-046 102851-048
4'-Bromo-3-(4-chlorophenyl)propiophenone ≥97%
4'-Bromo-3-(4-chlorophenyl)propiophenone

Hazard: Irritant

Formula: C₁₅H₁₂BrClO
MDL Number: MFCD03843467
CAS Number: 296271-45-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR